BRUTE® Vented Dustbins
Highly durable waste containers created with the toughest enviroments in mind.
- Intuitive design features: venting channels allow air flow around and below the bag, making for low force liner removal that is reduced by up to 50%; liner cinches prevent liners from falling into the bin, eliminating any need to tie knots
- Comfortable handling experience: reinforced rounded handles, with contoured base handles that assist with tipping
- Guaranteed durability: UV inhibitor ensures that BRUTE® containers never fade, crack, warp, or crush, even in the most extreme commercial settings
- Long-lasting: reinforced base means that the bin can be dragged over rough surfaces, extending the container's longevity in tough environments
- Food safe and compliant: all BRUTE® containers are certified EU food contact compliant, and also meet NSF 2, 21 approval
- Handles: contoured base handles improve grip and ergonomics, reducing strain and improving efficiency when emptying containers
- Strength: rim features integrated rib structure to increase strength and resist crushing
The BRUTE® waste container features innovative venting channels which make lifting out liners up to 50% easier. The proprietary design, constructed with the highest quality material plus a UV inhibitor, ensures long life in even the most extreme commercial environments. Guaranteed to never fade, warp, crack, or crush.