Adapters for Double Blood Bag Buckets for Thermo Scientific™ HAEMAFlex™ 12/16 Rotors
Supplier: Thermo Scientific
Certificates
About this item
Thermo Scientific blood banking centrifuges offer the flexibility to select higher capacity rotors or match existing workflows, with a choice of Thermo Scientific™ HAEMAFlex™ rotors and bucket systems.
- For high-throughput blood processing needs
- Double quint blood bags (XXL, XL or M size)
- Hook adapters for cord blood separations (XXL or M size)
For high-throughput blood processing needs, the HAEMAFlex 12 swinging bucket rotor provides 12×500 ml capacity or upgrade your centrifuge capacity to 16×500 ml blood bags with the HAEMAFlex 16 rotor and the same bucket selections.