Sterilin™ Borosilicate Glass Culture Tubes
Supplier: Sterilin
Certificates
About this item
Rely on chemical and temperature resistant borosilicate glass culture tubes in your tough applications. Thermo Scientific™ Sterilin™ Borosilicate glass culture tubes are available with flat or round bottom, threaded to accept a screw cap. Not available in America.
- High resistance to temperature and chemical attack
- Ideal for use with samples sensitive to leaching from plastic tubes
- Tube profile screw thread, culture
- Supplied without caps
- Autoclavable caps available separately
- Autoclavable at 121 °C
- Available in UK only