BASE Footwear Grand Canyon/Tulum Safety Shoes, Lace-Up, Low, S1P, Black
Supplier: Portwest
Certificates
About this item
Grand Canyon/Tulum low safety shoes in black feature a Microfibre + Hi-Tech fabric upper, SmellStop anti-bacterial lining, Fresh’n Flex midsole, SlimCap toe cap, and AirTech + TPU-skin sole.
- 200 joules resistant toe-cap
- Puncture-resistant sole
- Shock absorption in the heel region
- Anti-static footwear
- Electronic devices protection from electrostatic discharges
- Hydrocarbons-resistant
- Metal-free
Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces. Orthopaedic made to measure footbed, complying with european norms DGUV112-191.
Certifications: EN ISO 20345: 2011 S1P FO ESD SR; CEI EN 61340-4-3 S1P.