Order Entry
Finland
ContactUsLinkComponent
Chlorophyllin, coppered trisodium salt, powder
Chlorophyllin, coppered trisodium salt, powder
Catalog # SPCMC1443-12KGBL
Supplier:  Spectrum Chemical
CAS Number:  
Chlorophyllin, coppered trisodium salt, powder
Catalog # SPCMC1443-12KGBL
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly pail
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • Content of Chelated Copper:
    4.0% min.
  • Content of Total Copper:
    4.25 % min.
  • pH (1% in Water):
    9.0 - 10.9
  • Ash:
    max. 36 %
  • Arsenic:
    max. 3 ppm

Specification Test Results

About this item

 :
 :724.15 g/mol
Storage Temperature:Ambient
CAS Number:11006-34-1
Formula:C₃₄H₃₁CuN₄Na₃O₆
MDL Number:MFCD00012149