Order Entry
Finland
ContactUsLinkComponent
 
Irritant
Diazolidinyl urea ≥95%
Catalog # SPCMD2841-20KGBL
Supplier:  Spectrum Chemical
CAS Number:  
Irritant
Diazolidinyl urea ≥95%
Catalog # SPCMD2841-20KGBL
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Fiber drum
  • Pk:
    20 kg
  • Seq:
    4241

Specifications

Specification Test Results

  • Assay:
    Approx. 95%

Specification Test Results

About this item

 :
 :278.22 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:78491-02-8
Formula:C₈H₁₄N₄O₇
MDL Number:MFCD03547942