Order Entry
Finland
Orders LinkContactUsLinkComponent
D-ɑ-Tocopherol succinate ≥96.0% USP
D-ɑ-Tocopherol succinate ≥96.0% USP
D-ɑ-Tocopherol succinate ≥96.0% USP
D-ɑ-Tocopherol succinate ≥96.0% USP
RRR-Vitamin E Succinate, 1 mg = 0.81 alpha-tocopherol equivalents, USP.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
 :  Vitamin E Succinate, D-α-Tocopherol Succinate, 4-Oxo-4-(((R)-2,5,7,8-tetramethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-yl)oxy)butanoic acid
  •  :4345-03-3
  •  :C₃₃H₅₄O₅
  •  :MFCD00072055
  •  :530.79 g/mol
  •  :Ambient
  • Assay:
    96.0 - 102.0%
  • Acidity:
    To Pass Test
  • Identification:
    To Pass Test
Product Family Options
  • Pack type
    Pk
  • Glass, amber
    1 kg
    1 * 1 kg
    1 * 1 kg - 1 810,00 €
    1 810,00 €
    1 * 1 kg
       1 810,00 €
  • 25 kg
    1 * 25 kg
    1 * 25 kg - 8 040,00 €
    8 040,00 €
    1 * 25 kg
       8 040,00 €
Suositukset