Order Entry
Finland
ContactUsLinkComponent
 
trans-Cinnamaldehyde ≥98%, Reagent Grade
Catalog # SPCMCI110-200LTBL
Supplier:  Spectrum Chemical
CAS Number:  
trans-Cinnamaldehyde ≥98%, Reagent Grade
Catalog # SPCMCI110-200LTBL
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pk:
    200 L
  • Seq:
    5549

Specifications

Specification Test Results

  • Assay:
    98% Min.

Specification Test Results

About this item

 :
 :132.16 g/mol
Merck Index:12,02319
Boiling Pt:252 °C (1013 hPa)
Melting Pt:–8 °C
Density:1,05 g/cm³ (20 °C)
Flash Pt:138 °C
Storage Temperature:Refrigerator
CAS Number:14371-10-9
Formula:C₉H₈O
MDL Number:MFCD00007000