Order Entry
Finland
ContactUsLinkComponent
Tetrasodium diphosphate decahydrate ≥99%, crystals ACS
Irritant
Tetrasodium diphosphate decahydrate ≥99%, crystals ACS
Catalog # SPCMS1420-500GM
Supplier:  Spectrum Chemical
CAS Number:  
Irritant
Tetrasodium diphosphate decahydrate ≥99%, crystals ACS
Catalog # SPCMS1420-500GM
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay:
    99% Min.

Specification Test Results

About this item

 :
 :446.06 g/mol
Melting Pt:990 °C
Density:1,82 g/cm³ (25 °C)
EINECS:231-767-1
 :GHS07
Storage Temperature:Ambient
CAS Number:13472-36-1
Formula:Na₄P₂O₇·10H₂O
MDL Number:MFCD00149200