Anti-REG1A Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-6183R-A750
: Bioss
- Antibody type:Primary
- Antigen name:REG1A
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:101-166/166
- Form:Liquid
- Gene ID:5967
- Antigen synonyms:Pancreatic stone protein|PSP|REG1A|Lithostathine-1-alpha|Islet cells regeneration factor|P19|PTP|REG 1 beta|Regenerating islet-derived protein 1-alpha|ICRF|PSPS|Pancreatic thread protein|Pancreatic stone protein 2|REG1B|REG1A_HUMAN|PSPS2|Islet of Langerhans regenerating protein|Regenerating islet derived protein 1 beta|Regenerating protein I alpha|Lithostathine 1 beta|PSPS1|REG-1-alpha|REG|REGL
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:16 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human REG1A/REG1 alpha
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Reg protein is stimulated during the regeneration of pancreatic islets. In human, there are four REG family genes; REG 1 alpha, REG 1 beta, REG-related sequence (RS) and HIP/PAP. These Reg-related proteins are classified into four subfamilies according to their amino-acid sequences, but they share a similar structure and physiological function.
Type: Primary
Antigen: REG1A
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-166/166
Host: Rabbit
Isotype: IgG
Reactivity: