Order Entry
Finland
ContactUsLinkComponent
Anti-RASA3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-RASA3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-5954R-A750
Supplier:  Bioss
Anti-RASA3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-5954R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    RASA3
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    501-600/834
  • Form:
    Liquid
  • Gene ID:
    22821
  • Antigen synonyms:
    GAPIII|RASA3|GAP1IP4BP|GAP1(IP4BP)|Ras GTPase-activating protein 3|Ins P4-binding protein
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    91 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human RASA3
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Inhibitory regulator of the Ras-cyclic AMP pathway. Binds inositol tetrakisphosphate (IP4) with high affinity. Might be a specific IP4 receptor.

Type: Primary
Antigen: RASA3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 501-600/834
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat