Safety Waste Caps, Electrically Conductive
: S.C.A.T.
SCAT Safety Waste Caps, made of electrically conductive PTFE, are hermetically sealing caps for the safe disposal of your liquid HPLC waste (e.g. solvents, acids, alkalis).
- For the prevention of explosions due to static discharge
- Made of electrically conductive PTFE material
- Available in different thread sizes
The SCAT exhaust filter with three layers of activated carbon binds vapors. It thus additionally ensures safe pressure equalization and occupational safety.