Order Entry
Finland
ContactUsLinkComponent
Estrogen Receptor beta (ERβ) (Human) ELISA Kit
Estrogen Receptor beta (ERβ) (Human) ELISA Kit
Catalog # ABCAAB285292-96
Supplier:  Abcam
CAS Number:  
undefined
Estrogen Receptor beta (ERβ) (Human) ELISA Kit
Catalog # ABCAAB285292-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Human
  • Description:
    Estrogen Receptor beta (ERβ) (Human) ELISA kit

Specifications

About this item

An Sandwich ELISA kit to quantify human ER-beat in human serum, plasma and other biological fluids.

  • Easy, convenient and time-saving method to measure the level of ER-beta in human serum, plasma, tissue homogenates, culture supernatants and other biological fluids
  • Detection Range: 0,313 to 20 ng/ml
  • Sensitivity: <0,188 ng/ml

Nuclear hormone receptor. Binds estrogens with an affinity similar to that of ESR1, and activates expression of reporter genes containing estrogen response elements (ERE) in an estrogen-dependent manner. Isoform beta-cx lacks ligand binding ability and has no or only very low ere binding activity resulting in the loss of ligand-dependent transactivation ability. DNA-binding by ESR1 and ESR2 is rapidly lost at 37 degrees Celsius in the absence of ligand while in the presence of 17 beta-estradiol and 4-hydroxy-tamoxifen loss in DNA-binding at elevated temperature is more gradual. BioVision’s ER-beta ELISA kit is a sandwich ELISA assay for the quantitative measurement of human ER-beta in serum, plasma and cell culture supernatants. The density of colour is proportional to the amount of human ER-betacaptured from the samples.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.