ELISA Kit, Human Kininogen-1/KNG1 PicoKine
BSBTEK1154
:
- Pk:1 KIT
- Assay Type:Sandwich
- Primary antibody reactivity:Human
- Description:Human kininogen-1/KNG1 PicoKine; ELISA kit
Sandwich high sensitivity ELISA kit for quantitative detection of human kininogen-1/KNG1.
- Sensitivity, detect single digit picogram level of target protein
- Specificity, thoroughly tested for cross-reactivity with family members
- Stability, guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, on-demand technical support directly from ELISA experts
Detect human kininogen/KNG1 with <20 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Kininogen/KNG1 tissue specificity: secreted in plasma. T-kinin is detected in malignant ovarian, colon and breast carcinomas, but not in benign tumors.
96wells/kit, with removable strips.