Specifications
- Antibody type:Primary
- Antigen name:5HT1E Receptor/SR-1E
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Epitope:1-120/365
- Form:Liquid
- Antigen synonyms:Serotonin receptor 1E|S31|5 hydroxytryptamine serotonin receptor 1E|5HT1E_HUMAN|5 hydroxytryptamine 1E receptor|5-hydroxytryptamine receptor 1E|5 hydroxytryptamine serotonin receptor 1E G protein coupled|5-HT-1E|HTR1E
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:42 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human 5HT1E Receptor/SR-1E
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
The members of the G-protein-coupled receptor family are distinguished by their slow transmitting response to ligand binding. These seven transmembrane proteins include the adrenergic, serotonin and dopamine receptors. The effect of the Signalling molecule can be excitatory or inhibitory depending on the type of receptor to which it binds. b-adrenergic bound to adrenaline activates adenylyl cyclase, while a2-adrenergic receptor bound to adrenaline inhibits adenylyl cyclase. Like the a2-adrenergic receptor, serotonin receptor functions are also mediated by G proteins that inhibit the activity of adenylyl cyclase. The serotonin receptors have been classified into several categories, designated SR-17 (5HT17). Subtypes within the SR-1 group include SR-1A, -1B, -1D, -1E and -1F.
Type: Primary
Antigen: 5HT1E Receptor/SR-1E
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-120/365
Host: Rabbit
Isotype: IgG
Reactivity: