Anti-IL-23R Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
BOSSBS-1460R-FITC
: Bioss
- Antibody type:Primary
- Antigen name:IL-23R
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Dog,Mouse
- Western blot:Yes
- Epitope:401-550/629
- Form:Liquid
- Gene ID:149233
- Antigen synonyms:IL23R|Interleukin-23 receptor|IL-23R|IL-23 receptor
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:67 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human IL-23R
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Associates with IL12RB1 to form the interleukin-23 receptor. Binds IL23 and mediates T-cells, NK cells and possibly certain macrophage/myeloid cells stimulation probably through activation of the Jak-Stat signaling cascade. IL23 functions in innate and adaptive immunity and may participate in acute response to infection in peripheral tissues. IL23 may be responsible for autoimmune inflammatory diseases and be important for tumorigenesis.
Type: Primary
Antigen: IL-23R
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 401-550/629
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Dog