Anti-Aggrecan Core Protein 1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-1223R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Aggrecan core protein 1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rabbit,Human,Rat,Mouse
- Western blot:Yes
- Epitope:101-220/2415
- Form:Liquid
- Gene ID:176
- Antigen synonyms:Chondroitin sulfate proteoglycan 1|Cartilage-specific proteoglycan core protein|Aggrecan core protein|CSPCP|AGCAN|CSPG1|AGC1|SEDK|CSPGCP|MSK16|ACAN|Chondroitin sulfate proteoglycan core protein 1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:208/248 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Aggrecan core protein
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
This proteoglycan is a major component of extracellular matrix of cartilagenous tissues. A major function of this protein is to resist compression in cartilage. It binds avidly to hyaluronic acid via an N-terminal globular region.
Type: Primary
Antigen: Aggrecan core protein 1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-220/2415
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit