Anti-Versican Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-2533R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Versican
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Epitope:3301-3396/3396
- Form:Liquid
- Gene ID:1462
- Antigen synonyms:WGN1|PG-M|Chondroitin sulfate proteoglycan 2|Glial hyaluronate-binding protein|Large fibroblast proteoglycan|VCAN|GHAP|WGN|Chondroitin sulfate proteoglycan core protein 2|CSPG2|Versican core protein|ERVR
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:370 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Versican core protein
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
May play a role in intercellular signaling and in connecting cells with the extracellular matrix. May take part in the regulation of cell motility, growth and differentiation. Binds hyaluronic acid.
Type: Primary
Antigen: Versican
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 3301-3396/3396
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat