Anti-IKB epsilon (Ser22) Rabbit Polyclonal Antibody (ALEXA FLUOR® 488)
Catalog # BOSSBS-15582R-A488
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:IKB epsilon (Ser22)
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:IkB-E|MGC72568|NF kappa B inhibitor epsilon|NFkappaB inhibitor epsilon|Nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor epsilon|NF kappa BIE|IKBE_MOUSE|IkappaBepsilon|OTTHUMP00000016522|NF-kappa-B inhibitor epsilon|NFkappa BIE|Slc35b2|IkB E|NF-kappa-BIE|IkB-epsilon|solute carrier family 35, member B2|I kappa B epsilon|I-kappa-B-epsilon|IKBE|NFKBIE
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:39 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthesised phosphopeptide derived from human IKB epsilon around the phosphorylation site of Ser22
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
IKB epsilon (Ser22).
Type: Primary
Antigen: IKB epsilon (Ser22)
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 488
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat