About this item
For the quantification of PGIM in urine and other sample matrices.
- Measure PGIMs in urine
- Measure PGIM down to 120 pg/ml
- Convenient overnight format
Prostaglandin I (Prostaglandin I₂; PGI₂) is a potent vasodilator and inhibitor of platelet aggregation that is formed from arachidonic acid primarily in the vascular endothelium and renal cortex by sequential activities of cyclooxygenase (COX) and PGI₂ synthase. It is non-enzymatically hydrated to 6-keto PGF1α (t½ = 2 to 3 minutes), and then quickly converted to numerous downstream metabolites, collectively known as prostaglandin I metabolites or PGIM, (t½= 30 minutes). Although 6-keto PGF1α is commonly measured in plasma and urine as an estimate of PGI₂ synthesis, it should be noted that there may be more than one source of PGI₂ in these samples. For instance, urinary concentrations of 6-keto PGF1α are confounded by the fact that only a portion originates from plasma and the remainder is produced by the kidney. These problems are circumvented by measuring downstream PGIM rather than 6-keto PGF1α as an indicator of systemic PGI₂ production. Cayman's PGIM ELISA Kit is a competitive assay that can be used for quantification of PGIM in urine and other sample matrices. The assay has a range from 39 to 5000 pg/ml and a sensitivity (80% B/B₀) of approximately 120 pg/ml.
Contents: Antibody, tracer, standard, buffer, polysorbate 20, reagent, dyes, plate, and cover sheet.
Specifications
- Assay Type:Competitive
- Mid point:500 pg/ml
- Detection range:3,9 - 5000 pg/ml
- Sensitivity:120 pg/ml