GenFind V3 Reagent Kits
Catalog # BECKC34880
Supplier: Beckman Coulter
CAS Number:
Specifications
- Pk:1
- Sample type:Blood
- Description:GenFind V3 reagent kit, 50 preparations
- Format:Liquid
Specifications
About this item
This prep system kit isolates and purifies genomic DNA (gDNA) from whole blood. The kit is uses Solid Phase Reversible Immobilisation (SPRI) paramagnetic bead-based technology to effectively produce a high recovery of high quality DNA for downstream applications such as PCR.
- Automated available
Application: Genomic DNA extraction from whole blood.