Order Entry
Finland
ContactUsLinkComponent
 
Catalase Assay Kit
Catalog # CAYM707002-480
Supplier:  Cayman Chemical
CAS Number:  
undefined
Catalase Assay Kit
Catalog # CAYM707002-480
Supplier:  Cayman Chemical
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    Kit contains: Buffers, standard, control, Potassium Hydroxide, Hydrogen Peroxide; Chromogen, Potassium Periodate, and plate

Specifications

About this item

For the measurement of catalase activity in plasma, serum, erythrocyte lysates, tissue homogenates, and cell lysates.

  • Assay 40 samples in duplicate
  • Measure catalase activity down to 2 U/ml
  • Plate-based colorimetric measurement (540 nm)

Catalase is an ubiquitous antioxidant enzyme that is present in most aerobic cells. It is involved in the detoxification of H2O2, a ROS that is a toxic product of both normal aerobic metabolism and pathogenic ROS production. Cayman’s Catalase Assay utilises the peroxidatic function of catalase for determination of enzyme activity.