Order Entry
Finland
ContactUsLinkComponent
Arachidonic Acid (AA) ELISA Kit
Arachidonic Acid (AA) ELISA Kit
Catalog # ABCAAB287798-96
Supplier:  Abcam
CAS Number:  
undefined
Arachidonic Acid (AA) ELISA Kit
Catalog # ABCAAB287798-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay Type:
    Competitive
  • Description:
    Arachidonic acid ELISA kit
  • Tests per kit:
    100 assays

Specifications

About this item

A competitive ELISA kit for the quantitative measurement of arachidonic acid in biological fluids.

  • Detection range: 78,125 to 5000 ng/ml
  • Sensitivity: <69,74 ng/ml
  • Assay precision: Intra-assay: CV <8%; inter-assay: CV <10%
  • No significant cross-reactivity or interference was observed

Easy, convenient and time-saving method to measure the level of arachidonic acid in serum, plasma, tissue homogenates, culture supernatants and other biological fluids.

Arachidonic acid (AA, ARA) is a polyunsaturated omega-6 fatty acid 20:4(ω-6). It is structurally related to the saturated arachidic acid found in cupuaçu butter. It is a precursor to a large family of eicosanoids. Activator for several protein kinase C isotypes.

Arachidonic acid ELISA kit is a competitive ELISA assay for the quantitative measurement of arachidonic acid in serum, plasma and cell culture supernatants. The density of colour is proportional to the amount of arachidonic acid captured from the samples.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.