Decontamination Solution, BodyNeutrAll
PLUM4735
:
- Pk:1
- Description:Plum BodyNeutrAll, 5 L
- Seq:03
Mobile body shower with a highly effective decontamination solution for skin, surfaces and clothing. For fast and effective decontamination of bodies and surfaces in contact with corrosive and toxic hazardous substances such as strong acids and bases, oxidants, formaldehyde, phenols, organic solvents, hydrofluoric acid, and fluorides.
- Reduces strong oxidants (such as chlorine, hypochlorite, chlorine dioxide, chloramine, bromine, iodine, fluorine, hydrogen peroxide, peracetic acid) to compounds that are no longer active
- Binds aldehydes into imines, phenols into ammines, and heavy metals into ammine complexes
- Binds hydrofluoric acid and fluorides into sparingly soluble calcium fluoride
- Dissolves organic solvents and chemicals
- A red coloration of the sprayed area shows that oxidising agents have been inactivated
- Shelf life: 3 years from date of manufacture
600 ml – easy to use on smaller areas up to 6 m² or up to 3 persons.
100 ml – is pocket-sized to carry with you and covers approx. 1 m² or 1 arm's length.
5 L – suitable for larger areas up to 50 m² and covers several people.