Crucibles, stainless steel
Crucibles without lid made from stainless steel 18/10.
- Highly resistant to chemicals and reagents
- Withstand maximum temperature of 800 °C
- Autoclavable
: Lids need to be ordered separately.
Crucibles without lid made from stainless steel 18/10.