Order Entry
Finland
ContactUsLinkComponent
  116-0952
 :  DESCO
undefined
  116-0952
 :  DESCO
  • Colour:
    Yellow/black
  • Description:
    Caution label, English
  • Dimensions:
    25×50 mm
  • Pk:
    1 Roll
Caution labels made of white woodfree machine coated midgloss paper.
  • Marked with ESD susceptibility symbol
IEC 61340-5 requires all ESD sensitive devices and assemblies to be labelled with the 'susceptibility' symbol. It is intended to identify devices and assemblies that are susceptible to ESD. Mentioned on individual components and related documents to state "this device is static sensitive: do not touch without approrpiate precautions"
 : Compliant with IEC 61340-5-1.
 : Supplied in rolls of 1000 individual labels.