Order Entry
Finland
ContactUsLinkComponent
Recombinant Collagen V (from E. coli)
Recombinant Collagen V (from E. coli)
Catalog # NOVUNBP2-38162PEP
Supplier:  Novus Biologicals
CAS Number:  
Recombinant Collagen V (from E. coli)
Catalog # NOVUNBP2-38162PEP
Supplier:  Novus Biologicals
CAS Number:  

Specifications

  • Pk:
    0,1 mL
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Gene ID:
    COL5A1
  • Amino acid number:
    PVEAAKETTEVPEELTPTPTEAAPMPETSEGAGKEEDVGIGDYDYVPSEDYYTPSPYDDLTYGEGEENPDQPTDPGAGAEIPTSTADTSNSSNPAPPPGEGADDLEGEFTEETIRNLDENYYDPYYDPTSSPSEIGPGMPANQDTIYE
  • Protein synonyms:
    collagen alpha-1(V) chain|collagen, type V, alpha 1|alpha 1 type V collagen|EC 2.7.7.6|Collagen-5|EC 6.1.1
  • Protein/peptide name:
    Collagen
  • Purity:
    >80% by SDS-PAGE and Coomassie blue staining
  • Formulation:
    PBS and 1M Urea, pH 7.4. No Preservative

Specifications

About this item

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human COL5A1.

Type V collagen is a minor component of the connective tissue, although it is present in many different types of connective tissue. Patients with defects in the type V collagen (Ehlers-Danlos syndrome) have weakened connective tissue characterized by hyperstrechable joints and fragile, easily bruisable skin.