Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-3-methyl-5-nitrobenzoic acid
2-Bromo-3-methyl-5-nitrobenzoic acid ≥95%
2-Bromo-3-methyl-5-nitrobenzoic acid ≥95%
Irritant
2-Bromo-3-methyl-5-nitrobenzoic acid ≥95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Bromo-3-methyl-5-nitrobenzoic acid ≥95%
Supplier:
Apollo Scientific
About this item
CAS Number:
631911-95-0
:
Formula:
C₈H₆BrNO₄
MDL Number:
MFCD28128937
:
260.04 g/mol
:
GHS07