Welding gloves, EDGE® 48-216
Supplier: Ansell
Certificates
About this item
These protective gloves offer EN 12477 Type A protection for demanding welding and thermal job tasks, ranging from stick and MIG welding to torch cutting and grinding.
- Improved three-dimensional thumb design
- High-dexterity gloves offer tactile sensitivity for precise welding control
- Long cuff extends protection over the forearm
- Higher durability as they are made of premium split cowhide leather and have welted seams sewn with aramid
- Breathable cotton lining grants thermal insulation and extended comfort
Recommended for: Brazing, handling hot metal castings, grinding, welding, handling of metal sheets, cutting of dry, painted or galvanised pieces.
Certifications: EN 388:2016 (4133B), EN 407 (413X4X), EN 12477:2001 + A1:2005; Cat. III
Specifications
- Catalog No:
- ANSE48-216/10
- ANSE48-216/11
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Liner material:
- Cotton
- Cotton
- Material:
- Split cowhide
- Split cowhide
- Cuff style:
- Gauntlet
- Gauntlet
- EN norm:
- EN 388:2016 (4133B), EN 407 (413X4X), EN 12477:2001 + A1:2005
- EN 388:2016 (4133B), EN 407 (413X4X), EN 12477:2001 + A1:2005
- Sterility:
- Non sterile
- Non sterile