Order Entry
Finland
ContactUsLinkComponent
GOJO® HAND MEDIC® ADX-7™ Skin Conditioner Dispensers
GOJO® HAND MEDIC® ADX-7™ Skin Conditioner Dispensers
  115-2959
undefined
GOJO® HAND MEDIC® ADX-7™ Skin Conditioner Dispensers
  115-2959

 

  • Capacity:
    700 ml
  • Colour:
    Chrome/black
  • Description:
    GOJO® HAND MEDIC® ADX-7™ dispenser
  • Seq:
    02
  • Pk:
    1

 

 

Skin conditioner dispenser.

  • Compact design ideal for tight spaces
  • Large sight window for at-a-glance monitoring
  • ADA compliant for protrusion into walkways
  • Lifetime guarantee
  • Engineering grade plastics (ABS) construction for trouble-free performance
  • Multiple colors and finishes available

Dispenser is compatible with GOJO® HAND MEDIC® 685 ml refills.