Potassium lactate solution 60%
Glass, amber, 1 kg
Catalog # SPCMP1806-1KG
Supplier: Spectrum Chemical
Potassium lactate solution 60%
Catalog # SPCMP1806-1KG
Supplier: Spectrum Chemical
CAS Number: 996-31-6
Jump to:
Product Details & Documents
Potassium Lactate, 60 Percent Solution is used in personal care due to its high moisturizing properties as well as for its buffering capacity.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Synonyms: Potassium 2-hydroxypropanoate, Lactic acid, potassium salt
- CAS Number:996-31-6
- Formula:C₃H₅O₃K
- MDL Number:MFCD00036410
- Molecular Weight:128.168 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Glass, amber
- Pk:1 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



