Order Entry
Finland
ContactUsLinkComponent
Coliform PA broth dehydrated culture media
Coliform PA broth dehydrated culture media
Catalog # HIMEM1051-500G
Supplier:  HIMEDIA
CAS Number:  
Coliform PA broth dehydrated culture media
Catalog # HIMEM1051-500G
Supplier:  HIMEDIA
CAS Number:  
Specifications
  • Pk:
    500 g
  • Description:
    Coliform PA broth
Specifications
About this item
For the determination of the presence or absence of coliform bacteria in examination of pollution in treated water from water treatment plants or distribution systems.
  • Microbiological culture media
  • 500 g poly bottles