Order Entry
Finland
ContactUsLinkComponent
Ethyl acetate, CHROMASOLV™ for HPLC, Riedel-de Haën™
Ethyl acetate, CHROMASOLV™ for HPLC, Riedel-de Haën™
Catalog # HONC34858D-7L
   Safer Human and Environmental Health
CAS Number:  
Ethyl acetate, CHROMASOLV™ for HPLC, Riedel-de Haën™
Catalog # HONC34858D-7L
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Stainless steel can, returnable
  • Pk:
    7 L
  • Seq:
    5202
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

About this item

 :
 :88.11 g/mol
Boiling Pt:77 °C (1,013 hPa)
Melting Pt:–84 °C
Density:0.901 g/cm3
Flash Pt:–4 °C
Storage Temperature:Ambient
EINECS:205-500-4
UN:1173
ADR:3,II
CAS Number:141-78-6
Formula:CH₃COOC₂H₅
MDL Number:MFCD00009171