Order Entry
Finland
Orders LinkContactUsLinkComponent
Nebivolol, Reference Standard

Nebivolol, Reference Standard

Nebivolol Stereoisomer Mixture, USP reference standard, Vial, 15 mg
Catalog # 1457797.
Supplier:   USP
Nebivolol, Reference Standard
Catalog # 1457797.
Supplier:  USP
CAS Number:  
Nebivolol, Reference Standard

Nebivolol Stereoisomer Mixture, USP reference standard, Vial, 15 mg

CAS Number:  
Price
1 290,00 €
Log in for availability
1 * 15 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    15 mg
  • Standards:
    USP reference standard
  • Description:
    Nebivolol Stereoisomer Mixture
  • UN No.:
    3077
  • Cat. No.:
    1457797.
  • Description:
    Nebivolol Stereoisomer Mixture

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.