Order Entry
Finland
Orders LinkContactUsLinkComponent
Raspberry Alcohol, Reference Standard

Raspberry Alcohol, Reference Standard

Raspberry Alcohol, USP reference standard, Vial, 50 mg
Catalog # 1598802.
Supplier:   USP
Raspberry Alcohol, Reference Standard
Catalog # 1598802.
Supplier:  USP
CAS Number:  
Raspberry Alcohol, Reference Standard

Raspberry Alcohol, USP reference standard, Vial, 50 mg

CAS Number:  
Price
375,00 €
Log in for availability
1 * 50 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    50 mg
  • Standards:
    USP reference standard
  • Description:
    Raspberry Alcohol
  • Cat. No.:
    1598802.
  • Description:
    Raspberry Alcohol
  • Synonym(s):
    4-(3-Hydroxybutyl)phenol
  • CAS No.:
    69617-84-1

Specifications