Polyethylene Glycol, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
10 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
Certificates
Search by Lot NumberSpecifications for Product Family
Specifications
Specifications
- Catalog No:
- 1546401.
- 1546423.
- 1546445.
- 1546467.
- 1546489.
- 1546525.
- 1546547.
- 1546569.
- 1546580.
- 1546605.
- Description:
- Polyethylene Glycol 200
- Polyethylene Glycol 300
- Polyethylene Glycol 400
- Polyethylene Glycol 600
- Polyethylene Glycol 1000
- Polyethylene Glycol 3000
- Polyethylene Glycol 3350
- Polyethylene Glycol 4000
- Polyethylene Glycol 6000
- Polyethylene Glycol 8000
- Synonym(s):
- PEG
- PEG
- PEG
- PEG
- PEG
- PEG
- PEG
- PEG
- PEG
- CAS No.:
- 25322-68-3
- 25322-68-3
- 25322-68-3
- 25322-68-3
- 25322-68-3
- 25322-68-3
- 25322-68-3
- 25322-68-3
- 25322-68-3
- 25322-68-3
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1546401.Polyethylene Glycol 200Specifications:
More
Specifications
Cat. No.1546401.DescriptionPolyethylene Glycol 200Synonym(s)PEGCAS No.25322-68-3 - 1546423.Polyethylene Glycol 300Specifications:
More
Specifications
Cat. No.1546423.DescriptionPolyethylene Glycol 300Synonym(s)PEGCAS No.25322-68-3 - 1546445.Polyethylene Glycol 400Specifications:
More
Specifications
Cat. No.1546445.DescriptionPolyethylene Glycol 400Synonym(s)PEGCAS No.25322-68-3 - 1546467.Polyethylene Glycol 600Specifications:
More
Specifications
Cat. No.1546467.DescriptionPolyethylene Glycol 600Synonym(s)PEGCAS No.25322-68-3 - 1546489.Polyethylene Glycol 1000Specifications:
More
Specifications
Cat. No.1546489.DescriptionPolyethylene Glycol 1000Synonym(s)PEGCAS No.25322-68-3 - 1546525.Polyethylene Glycol 3000Specifications:
More
Specifications
Cat. No.1546525.DescriptionPolyethylene Glycol 3000Synonym(s)PEGCAS No.25322-68-3 - 1546547.Polyethylene Glycol 3350Specifications:
More
Specifications
Cat. No.1546547.DescriptionPolyethylene Glycol 3350Synonym(s)PEGCAS No.25322-68-3 - 1546569.Polyethylene Glycol 4000Specifications:
More
Specifications
Cat. No.1546569.DescriptionPolyethylene Glycol 4000Synonym(s)CAS No.25322-68-3 - 1546580.Polyethylene Glycol 6000Specifications:
More
Specifications
Cat. No.1546580.DescriptionPolyethylene Glycol 6000Synonym(s)PEGCAS No.25322-68-3 - 1546605.Polyethylene Glycol 8000Specifications:
More
Specifications
Cat. No.1546605.DescriptionPolyethylene Glycol 8000Synonym(s)PEGCAS No.25322-68-3



