Perphenazine, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
3 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1511203.
- 1511000.
- 1511021.
- Description:
- Perphenazine Sulfoxide
- Perphenazine
- Perphenazine Related Compound B
- Synonym(s):
- 2-{4-[3-(10H-phenothiazin-10-yl)propyl]piperazin-1-yl}ethanol
- CAS No.:
- 10078-25-8
- 58-39-9
- 3533-97-9
- UN No.:
- 2811
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1511203.Perphenazine SulfoxideSpecifications:
More
Specifications
Cat. No.1511203.DescriptionPerphenazine SulfoxideSynonym(s)CAS No.10078-25-8UN No. - 1511000.PerphenazineSpecifications:
More
Specifications
Cat. No.1511000.DescriptionPerphenazineSynonym(s)CAS No.58-39-9UN No.2811 - 1511021.Perphenazine Related Compound BPerphenazine Related Compound BUSP reference standardVial15 mgLog in for availability1 * 15 mg1 210,00 €Specifications:
More
Specifications
Cat. No.1511021.DescriptionPerphenazine Related Compound BSynonym(s)2-{4-[3-(10H-phenothiazin-10-yl)propyl]piperazin-1-yl}ethanolCAS No.3533-97-9UN No.



