Order Entry
Finland
Orders LinkContactUsLinkComponent
Everolimus, Reference Standards

Everolimus, Reference Standards

Supplier:   USP
Everolimus, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Everolimus System Suitability Mixture: This is a mixture of everolimus, everolimus oxepane isomer and 9-O-hydroxyethyl sirolimus

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1268976.
  • 1268987.
  • Description:
  • Everolimus
  • Everolimus System Suitability Mixture
  • CAS No.:
  • 159351-69-6
  • UN No.:
  • 3077
  • 3077

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1268976.
    Everolimus
    Everolimus
    USP reference standard
    Vial
    300 mg
    Log in for availability
    Log in for availability
    1 * 300 mg
    1 * 300 mg - 2 690,00 €
    2 690,00 €
    1 * 300 mg
       2 690,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1268976.
    Description
    Everolimus
    CAS No.
    159351-69-6
    UN No.
    3077
  • 1268987.
    Everolimus System Suitability Mixture
    Everolimus System Suitability Mixture
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 2 890,00 €
    2 890,00 €
    1 * 25 mg
       2 890,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1268987.
    Description
    Everolimus System Suitability Mixture
    CAS No.
    UN No.
    3077
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.