Order Entry
Finland
Orders LinkContactUsLinkComponent
Digoxin, Reference Standard

Digoxin, Reference Standard

Digoxin, USP reference standard, Vial, 250 mg
Catalog # 1200000.
Supplier:   USP
Digoxin, Reference Standard
Catalog # 1200000.
Supplier:  USP
CAS Number:  
Digoxin, Reference Standard

Digoxin, USP reference standard, Vial, 250 mg

CAS Number:  
Price
416,00 €
Log in for availability
1 * 250 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    250 mg
  • Standards:
    USP reference standard
  • Description:
    Digoxin
  • UN No.:
    2811
  • Cat. No.:
    1200000.
  • Description:
    Digoxin
  • CAS No.:
    20830-75-5

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.