PFA tongs
Catalog # SAVI700-850
Supplier: SAVILLEX
Specifications
- Description:PFA tongs
- Pk:1
Specifications
About this item
Adjustable PFA tongs are moulded from the highest purity PFA resin for ultratrace metals use.
Service temperature range is –200 to +260 °C (–328 to +500 °F).
Ideal for transferring ICP glassware during cleaning. Tongs are 28 cm (11") long with three adjustable width positions.