Order Entry
Finland
ContactUsLinkComponent
DST (Disuccinimidyl tartrate), Pierce™
DST (Disuccinimidyl tartrate), Pierce™
Catalog # PIER20589
CAS Number:  
undefined
DST (Disuccinimidyl tartrate), Pierce™
Catalog # PIER20589
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

Specification Test Results

  • Product Line:
    Pierce™
  • Labeling Method:
    Chemical Labeling
  • Crosslinker Type:
    Homobifunctional
  • Reactive Moiety:
    NHS Ester
  • Spacer:
    Cleavable, Short
  • Form:
    Powder
  • Quantity:
    50 mg
  • Type:
    Crosslinker
  • Solubility:
    DMF, DMSO

Specification Test Results

About this item

 :
 :344.231 g/mol
Storage Temperature:Refrigerator
CAS Number:77658-91-4
 :MFCD00049060
Formula:C₁₂H₁₂N₂O₁₀
MDL Number:MFCD03788224