DST (Disuccinimidyl tartrate)
BC05
:
- Pk:1 g
- Seq:3047
- Spacer Arm (Å):6.4
- Reactive Toward:Primary Amines
- Membrane Permeable:Yes
- Water Soluble:No
- Cleavable/ Reversible:Oxidizing Agents (Periodate)
:
:344.231 g/mol
:77658-91-4
:MFCD00049060
:C₁₂H₁₂N₂O₁₀
:MFCD03788224