UA labels 'Clean'
Catalog # 300-0554
Supplier: Sampling Systems
Specifications
- Description:UA labels 'Clean'
- Length:95 mm
- Packed:500/roll
- Pk:1
- Width:95 mm
Specifications
About this item
Ultra-adhesive labels offer a wide range of uses.
- Incorporates a write-on paper front
- High strength, ultra-adhesive for a range of surfaces
- Perfect for dusty surfaces
- Ideal for sealing holes in bags
Ultra-adhesive labels provide a stronger hermetic barrier and can be used for a wide range of applications. The high strength adhesive used means these labels will even stick on very dusty surfaces.