Order Entry
Finland
ContactUsLinkComponent
Sulpho-NHS (N-Hydroxysulphosuccinimide sodium salt), No-Weigh™ Format, Pierce™
Sulpho-NHS (N-Hydroxysulphosuccinimide sodium salt), No-Weigh™ Format, Pierce™
Catalog # PIERA39269
CAS Number:  
Sulpho-NHS (N-Hydroxysulphosuccinimide sodium salt), No-Weigh™ Format, Pierce™
Catalog # PIERA39269
CAS Number:  

Specifications

  • Pk:
    20 mg
  • Seq:
    2305
  • Pk Info:
    10 x 2 mg

Specifications

Specification Test Results

  • Spacer Arm Length:
    Short

Specification Test Results

About this item

 :
 :217.13 g/mol
Storage Temperature:Refrigerator
CAS Number:106627-54-7
Formula:C₄H₄NNaO₆S
MDL Number:MFCD00043100