EMCH.TFA (E-Maleimidocaproic acid hydrazine trifluoroacetic acid salt) ≥90% (by NMR), Pierce™
Catalog # PIER22106
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
Specification Test Results
- Product Line:Pierce™
- Labeling Method:Chemical Labeling
- Crosslinker Type:Heterobifunctional
- Reactive Moiety:Amine, Hydrazide, Maleimide
- Spacer:Mid-Length
- Form:Solid
- Quantity:50 mg
- Type:Crosslinker
- Solubility:Water
Specification Test Results
About this item
:
:339.27 g/mol
Melting Pt:99…110°C
Storage Temperature:Refrigerator
CAS Number:151038-94-7
:175290-73-0
Formula:C₁₀H₁₅N₃O₃·C₂HF₃O₂
MDL Number:MFCD07357647