Heavy-duty hand reel
SAMP5070W-01
- Description:Heavy-duty hand reel
- Material:PP
- Pk:1
Essential for sampling with Sub Samplers and bailers.
- High capacity reel
- Built-in lcoking mechanism and hanging hooks
A popular range of hand reels and cables are essential when sampling with Sub Samplers and bailers.