AseptiQuik® X connectors
Supplier: CPC COLDER
Certificates
About this item
AseptiQuik® X large format 1" connectors provide quick and easy sterile connections for high flow applications, even in non-sterile environments. The 'Twist-Pull-Twist' design enables users to quickly transfer large volumes of media easily with less risk of operator error.
- Operating temperature: 4 to 40 °C
- Ensure simultaneous and secure removal of both membranes
- Audible confirmation of completed assembly steps
- Open access through multiple supply chain partners
- Secures final connection preventing disassembly
Biopharmaceutical manufacturers can now make 1" sterile connections with the quality and market availability they expect from the leader in single-use connection technology.
Certifications: USP Class VI, ADCF Certified.
Specifications
- Catalog No:
- CPCCAQX44024HT
- CPCCAQX33024HT
- Max. operating pressure:
- Vacuum...4,1 bar
- Vacuum...4,1 bar