
Ellagic acid dihydrate ≥98% (by HPLC), Calbiochem®, Sigma-Aldrich®
324683-500
:
- Pack type:Glass bottle
- Pk:500 mg
- Seq:2725
:
:302.19 g/mol
:M-Clarity™ quality level = MQ100
:207-508-3
:GHS07
:Ambient
:476-66-4
:C₁₄H₆O₈·2H₂O
:MFCD00006914