Order Entry
Finland
ContactUsLinkComponent
Anti-CD86 Rat Antibody [clone: GL-1 (GL1)]
Anti-CD86 Rat Antibody [clone: GL-1 (GL1)]
Catalog # 40-0862-U100
Supplier:  Tonbo Biosciences
undefined
Anti-CD86 Rat Antibody [clone: GL-1 (GL1)]
Catalog # 40-0862-U100
Supplier:  Tonbo Biosciences

Specifications

  • Antigen name:
    CD86 molecule
  • Antigen symbol:
    CD86
  • Clone:
    GL-1 (GL1)
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Mouse
  • Gene ID:
    12524
  • Antigen synonyms:
    CLS1|B70|B7.2|Ly-58|Cd28l2|ETC-1|MB7|TS/A-2|B7-2|B7|MB7-2|Ly58
  • Storage temperature:
    Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Tested applications:
    Functional Assay, Immunofluorescence Microscopy
  • Pk:
    100 µG

Specifications

About this item

The GL-1 antibody reacts with mouse CD86, also known as B7-2, an 80 kDa cell surface protein which is a ligand for CD28, a co-stimulatory receptor for the T cell receptor (TCR). CD28 can also bind a second B7 ligand known as CD80 (B7-1). Both CD80 and CD86 are expressed on activated B cells and antigen-presenting cells. These ligands trigger CD28 signaling in concert with TCR activation to drive T cell proliferation, induce high-level expression of IL-2, impart resistance to apoptosis, and enhance T cell cytotoxicity. The interaction / co-stimulatory signaling between the B7 ligands and CD28 provides crucial communication between T cells and B cells or APCs to coordinate the adaptive immune response. 

The GL-1 antibody may be used as a marker for CD86 expression on B cells, macrophages, and dendritic cells.

Type:
Antigen: CD86
Clonality:
Clone: GL-1 (GL1)
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: