Order Entry
Finland
ContactUsLinkComponent
iNOS overexpression lysate (adult normal)
iNOS overexpression lysate (adult normal)
Catalog # NOVUNBL1-13721
Supplier:  Novus Biologicals
CAS Number:  
iNOS overexpression lysate (adult normal)
Catalog # NOVUNBL1-13721
Supplier:  Novus Biologicals
CAS Number:  

Specifications

  • Cell Format:
    Cryopreserved
  • Pk:
    0,1 mg
  • Tested applications:
    WB

Specifications

About this item

iNOS Overexpression Lysate (Adult Normal)

Tested Applications: Western Blot.